Learn More
Diethyl ketomalonate, 95%, Thermo Scientific Chemicals
$80.37 - $902.47
Chemical Identifiers
CAS | 609-09-6 |
---|---|
Molecular Formula | C7H10O5 |
Molecular Weight (g/mol) | 174.152 |
MDL Number | MFCD00009121 |
InChI Key | DBKKFIIYQGGHJO-UHFFFAOYSA-N |
Synonym | diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester |
PubChem CID | 69105 |
IUPAC Name | diethyl 2-oxopropanedioate |
SMILES | CCOC(=O)C(=O)C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1277106
|
Thermo Scientific Chemicals
A1277106 |
5 g |
Each for $80.37
|
|
|||||
AAA1277114
|
Thermo Scientific Chemicals
A1277114 |
25 g |
Each for $255.81
|
|
|||||
AAA1277122
|
Thermo Scientific Chemicals
A1277122 |
100 g |
Each for $902.47
|
|
|||||
Description
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.
Solubility
Fully miscible with water.
Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Chemical Identifiers
609-09-6 | |
174.152 | |
DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
69105 | |
CCOC(=O)C(=O)C(=O)OCC |
C7H10O5 | |
MFCD00009121 | |
diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
diethyl 2-oxopropanedioate |
Specifications
609-09-6 | |
1.142 | |
>110°C (230°F) | |
1.415 | |
5 g | |
Moisture sensitive | |
diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
diethyl 2-oxopropanedioate | |
69105 | |
95% |
∼-70°C | |
208°C to 210°C | |
C7H10O5 | |
MFCD00009121 | |
971873 | |
14,5913 | |
Fully miscible with water. | |
CCOC(=O)C(=O)C(=O)OCC | |
174.152 | |
174.15 | |
Diethyl ketomalonate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-176-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only