missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl maleate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114442500

 View more versions of this product

Catalog No. AC114442500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl maleate
141-05-9
1.0600g/mL
93°C
96% min. (GC)
C8H12O4
C2H5O2CCH=CHCO2C2H5
250 g
1.06
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
IEPRKVQEAMIZSS-WAYWQWQTSA-N
diethyl (Z)-but-2-enedioate
5271566
172.18
Liquid
ca.90%
-10.0°C
225.0°C
Authentic
Glass bottle
1.4380 to 1.4420
MFCD00009191
02,751
15,3139
Solubility in water: insoluble. Other solubilities: readily soluble in alcohol, diethyl ether
CCOC(=O)C=CC(=O)OCC
172.18
CHEBI:68508
97%

Chemical Identifiers

141-05-9
MFCD00009191
diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
CHEBI:68508
CCOC(=O)C=CC(=O)OCC
172.18
IEPRKVQEAMIZSS-WAYWQWQTSA-N
5271566
diethyl (Z)-but-2-enedioate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-451-9

RTECSNumber : ON1225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only