missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl malonate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114450250

 View more versions of this product

Catalog No. AC114450250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl malonate
-50.0°C
199.0°C
Authentic
Glass bottle
1.4130 to 1.4150
25 g
10,182; 13,69
15,3878
Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone
CCOC(=O)CC(=O)OCC
160.17
2.14 mPa.s (10°C)
99+%
105-53-3
1.0550g/mL
93°C
99% min. (GC)
C7H12O4
C2H5O2CCH2CO2C2H5
02,573
1.055
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
diethyl propanedioate
7761
160.17
Liquid

Chemical Identifiers

105-53-3
160.17
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
diethyl propanedioate
C7H12O4
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
7761
CCOC(=O)CC(=O)OCC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-305-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only