Learn More
Diethyl phenylthiomethylphosphonate, 96%, Thermo Scientific Chemicals
$132.00 - $508.36
Chemical Identifiers
CAS | 38066-16-9 |
---|---|
Molecular Formula | C11H17O3PS |
Molecular Weight (g/mol) | 260.288 |
MDL Number | MFCD00015130 |
InChI Key | FBUXEPJJFVDUFE-UHFFFAOYSA-N |
Synonym | diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate # |
PubChem CID | 99227 |
IUPAC Name | diethoxyphosphorylmethylsulfanylbenzene |
SMILES | CCOP(=O)(CSC1=CC=CC=C1)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1366009
|
Thermo Scientific Chemicals
A1366009 |
10 g |
Each for $132.00
|
|
|||||
AAA1366018
|
Thermo Scientific Chemicals
A1366018 |
50 g |
Each for $508.36
|
|
|||||
Description
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
38066-16-9 | |
260.288 | |
FBUXEPJJFVDUFE-UHFFFAOYSA-N | |
99227 | |
CCOP(=O)(CSC1=CC=CC=C1)OCC |
C11H17O3PS | |
MFCD00015130 | |
diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate # | |
diethoxyphosphorylmethylsulfanylbenzene |
Specifications
38066-16-9 | |
146°C to 148°C (0.5 mmHg) | |
1.534 | |
10 g | |
diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate # | |
FBUXEPJJFVDUFE-UHFFFAOYSA-N | |
diethoxyphosphorylmethylsulfanylbenzene | |
99227 | |
96% |
1.174 | |
C11H17O3PS | |
MFCD00015130 | |
1246215 | |
Difficult to mix. | |
CCOP(=O)(CSC1=CC=CC=C1)OCC | |
260.288 | |
260.29 | |
Diethyl phenylthiomethylphosphonate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 253-770-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only