missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl Phosphoramidate 98.0+%, TCI America™
$64.70 - $214.02
Chemical Identifiers
CAS | 1068-21-9 |
---|---|
Molecular Formula | C4H12NO3P |
Molecular Weight (g/mol) | 153.118 |
MDL Number | MFCD00015676 |
InChI Key | MCQILDHFZKTBOD-UHFFFAOYSA-N |
Synonym | Diethyl Amidophosphate, Diethyl Phosphoryl Amide |
PubChem CID | 70605 |
IUPAC Name | [amino(ethoxy)phosphoryl]oxyethane |
SMILES | CCOP(=O)(N)OCC |
Chemical Identifiers
1068-21-9 | |
153.118 | |
MCQILDHFZKTBOD-UHFFFAOYSA-N | |
70605 | |
CCOP(=O)(N)OCC |
C4H12NO3P | |
MFCD00015676 | |
Diethyl Amidophosphate, Diethyl Phosphoryl Amide | |
[amino(ethoxy)phosphoryl]oxyethane |
Specifications
1068-21-9 | |
White-Yellow | |
C4H12NO3P | |
5 g | |
MCQILDHFZKTBOD-UHFFFAOYSA-N | |
[amino(ethoxy)phosphoryl]oxyethane | |
70605 | |
≥98.0% (GC) | |
Diethyl Phosphoramidate |
52°C | |
140°C | |
MFCD00015676 | |
Diethyl Amidophosphate, Diethyl Phosphoryl Amide | |
CCOP(=O)(N)OCC | |
153.118 | |
153.12 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1992
TSCA : No