missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrazole-3,5-dicarboxylate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 37687-24-4
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.21
MDL Number MFCD00152167
InChI Key MBWXLICVQZUJOW-UHFFFAOYSA-N
Synonym diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
PubChem CID 142184
IUPAC Name diethyl 1H-pyrazole-3,5-dicarboxylate
SMILES CCOC(=O)C1=CC(=NN1)C(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296520050
SDS View Documents
Thermo Scientific Chemicals
296520050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

37687-24-4
212.21
MBWXLICVQZUJOW-UHFFFAOYSA-N
142184
CCOC(=O)C1=CC(=NN1)C(=O)OCC
C9H12N2O4
MFCD00152167
diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
diethyl 1H-pyrazole-3,5-dicarboxylate

Specifications

37687-24-4
98.5% min. (GC)
C9H12N2O4
5g
MBWXLICVQZUJOW-UHFFFAOYSA-N
diethyl 1H-pyrazole-3,5-dicarboxylate
142184
99%
Authentic
Glass bottle
MFCD00152167
diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
CCOC(=O)C1=CC(=NN1)C(=O)OCC
212.21
212.21
Diethyl pyrazole-3,5-dicarboxylate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only