missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 381161000

 View more versions of this product

Catalog No. AC381161000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Diethylaluminium iodide
2040-00-8,108-88-3
AcroSeal™ Glass Bottle
(C2H5)2AlI
100 mL
Solubility in water: reacts
CC[Al](CC)I
212.01
212.01
0.8M Solution in Toluene
Colorless
C4H10AlI
MFCD00001025
aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali
PPQUYYAZSOKTQD-UHFFFAOYSA-M
diethyl(iodo)alumane
74874
Liquid

Chemical Identifiers

2040-00-8
212.01
PPQUYYAZSOKTQD-UHFFFAOYSA-M
74874
CC[Al](CC)I
C4H10AlI
MFCD00001025
aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali
diethyl(iodo)alumane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Reacts violently with water.

GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 218-032-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only