missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylamine, 99.5%, extra pure, redistilled, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 378371000

 View more versions of this product

Catalog No. AC378371000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylamine
-50.0°C
0.7100g/mL
−23°C
99.4% min. (GC)
C4H11N
(C2H5)2NH
100 mL
0.71
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
HPNMFZURTQLUMO-UHFFFAOYSA-N
N-ethylethanamine
8021
CHEBI:85259
99.5%
Liquid
109-89-7
Colorless
55.0°C
Authentic
Glass bottle
1.3840 to 1.3860
MFCD00009032
1154
15,3122
Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic,solvents
CCNCC
73.14
0.34 mPa.s (25°C)
73.13
Extra Pure

Chemical Identifiers

109-89-7
73.14
HPNMFZURTQLUMO-UHFFFAOYSA-N
8021
N-ethylethanamine
C4H11N
MFCD00009032
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
CHEBI:85259
CCNCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-716-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only