missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethylamine Acetate 97.0+%, TCI America™
Supplier: TCI America D135225G
Specifications
Diethylamine Acetate | |
47°C | |
C6H15NO2 | |
25 g | |
CCNCC.CC(=O)O | |
133.191 | |
133.19 | |
Crystalline Powder |
20726-63-0 | |
White-Yellow | |
MFCD00060243 | |
UEHUZQKLOWYOMO-UHFFFAOYSA-N | |
acetic acid;N-ethylethanamine | |
167925 | |
≥97.0% (T) |
Chemical Identifiers
20726-63-0 | |
133.191 | |
UEHUZQKLOWYOMO-UHFFFAOYSA-N | |
acetic acid;N-ethylethanamine |
C6H15NO2 | |
MFCD00060243 | |
167925 | |
CCNCC.CC(=O)O |
Safety and Handling
EINECSNumber : (2)-0135
TSCA : Yes