missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 155891000

 View more versions of this product

Catalog No. AC155891000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylamine hydrochloride
660-68-4
White
1% max.
99%
C4H11N·HCl
MFCD00012499
04,95
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
HDITUCONWLWUJR-UHFFFAOYSA-N
N-ethylethanamine;hydrochloride
10197650
99%
99%
227.0°C to 230.0°C
320.0°C to 330.0°C
Authentic
Plastic bottle
(C2H5)2NH·HCl
100 g
15,3122
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
CCNCC.Cl
109.6
109.6
Crystalline Powder or Crystals

Chemical Identifiers

660-68-4
HDITUCONWLWUJR-UHFFFAOYSA-N
10197650
CCNCC.Cl
MFCD00012499
diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
N-ethylethanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-541-9

RTECSNumber : IA3084000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only