Learn More
Diethylcarbamyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1570122
Description
It is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the production of pharma ingredients. It is a reagent in the synthesis of S-Methyl-N,N-diethylthiocarbamate Sulfoxide, which is an oxygenated metabolite of Disulfiram (Antabuse) that is capable of in vitro inactivation of liver mitochondrial aldehyde dehydrogenase.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications
-32°C | |
88-10-8 | |
C5H10ClNO | |
MFCD00000636 | |
506687 | |
1.451 | |
OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
N,N-diethylcarbamoyl chloride | |
6916 | |
98% | |
100 g | |
Moisture sensitive |
Diethylcarbamyl chloride | |
186°C | |
(CH3CH2)2NCOCl | |
UN3265 | |
diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride | |
Reacts with water. | |
CCN(CC)C(=O)Cl | |
135.591 | |
135.59 | |
1.07 | |
76°C (168°F) |
Chemical Identifiers
88-10-8 | |
135.591 | |
OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
6916 | |
CCN(CC)C(=O)Cl |
C5H10ClNO | |
MFCD00000636 | |
diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride | |
N,N-diethylcarbamoyl chloride |
Safety and Handling
GHS H Statement
H351-H302-H332-H315-H319-H335-H227
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P374-P501c
H227-H302+H332-H315-H319-H335-H351
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 201-798-5
RTECSNumber : FD4025000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only