missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylene glycol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A147280C

 View more versions of this product

Catalog No. AAA147280C

Explore available promotions

Description

Description

Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

Solubility
Miscible with water, alcohol, ether, acetone, and ethylene glycol.

Notes
Hygroscopic. Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and zinc.
Specifications

Specifications

Diethylene glycol
-10°C
245°C to 246°C
99%
C4H10O3
(HOCH2CH2)2O
10,000 g
Hygroscopic
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
MTHSVFCYNBDYFN-UHFFFAOYSA-N
2-(2-hydroxyethoxy)ethanol
8117
106.12
Liquid
111-46-6
1.118
143°C (289°F)
Nearly Odorless
1.447
MFCD00002882
969209
14,3119
Miscible with water,alcohol,ether,acetone,and ethylene glycol.
OCCOCCO
106.12
CHEBI:46807
99%

Chemical Identifiers

111-46-6
106.12
MTHSVFCYNBDYFN-UHFFFAOYSA-N
8117
OCCOCCO
C4H10O3
MFCD00002882
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
CHEBI:46807
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P260-P264b-P270-P301+P312-P314-P330-P501c

H302-H373

EINECSNumber : 203-872-2

RTECSNumber : ID5950000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only