Learn More
Diethylene glycol diacrylate, tech. 75%, stab.
CAS: 4074-88-8 | C10H14O5 | 214.22 g/mol
$49.78 - $112.26
Chemical Identifiers
| CAS | 4074-88-8 |
|---|---|
| Molecular Formula | C10H14O5 |
| Molecular Weight (g/mol) | 214.22 |
| MDL Number | MFCD00014939 |
| InChI Key | LEJBBGNFPAFPKQ-UHFFFAOYSA-N |
| Synonym | diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate |
| PubChem CID | 19996 |
| IUPAC Name | 2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate |
| SMILES | C=CC(=O)OCCOCCOC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1068214
|
Thermo Scientific Chemicals
L1068214 |
25 g |
N/A
|
|
|||||
|
AAL1068222
|
Thermo Scientific Chemicals
L1068222 |
100 g |
N/A
|
|
|||||
Description
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.
Solubility
Slightly soluble in water (1-5g/100ml at 18°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 4074-88-8 | |
| 214.22 | |
| LEJBBGNFPAFPKQ-UHFFFAOYSA-N | |
| 19996 | |
| C=CC(=O)OCCOCCOC(=O)C=C |
| C10H14O5 | |
| MFCD00014939 | |
| diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate | |
| 2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate |
Specifications
| 4074-88-8 | |
| 94°C (0.2 mmHg) | |
| C10H14O5 | |
| MFCD00014939 | |
| UN2810 | |
| diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate | |
| LEJBBGNFPAFPKQ-UHFFFAOYSA-N | |
| 2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate | |
| 19996 | |
| 75% | |
| Diethylene glycol diacrylate, stabilized |
| 1.118 | |
| >110°C (230°F) | |
| 1.462 | |
| 25 g | |
| 1953711 | |
| Slightly soluble in water (1-5g/100ml at 18°C). | |
| C=CC(=O)OCCOCCOC(=O)C=C | |
| 214.22 | |
| 214.22 | |
| Technical |
Safety and Handling
GHS H Statement
H311-H315-H319-H317
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P312-P333+P313-P361-P363-P501c
H311-H315-H317-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 223-791-6
RTECSNumber : AS9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only