missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Dibenzoate 97.0+%, TCI America™
$78.29 - $428.73
Chemical Identifiers
| CAS | 120-55-8 |
|---|---|
| Molecular Formula | C18H18O5 |
| Molecular Weight (g/mol) | 314.34 |
| MDL Number | MFCD00020679 |
| InChI Key | NXQMCAOPTPLPRL-UHFFFAOYSA-N |
| PubChem CID | 8437 |
| IUPAC Name | 2-[2-(benzoyloxy)ethoxy]ethyl benzoate |
| SMILES | O=C(OCCOCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D152225ML
|
TCI America
D152225ML |
25 mL |
Each for $78.29
|
|
|||||
|
D1522500ML
|
TCI America
D1522500ML |
500 mL |
Each for $428.73
|
|
|||||
Chemical Identifiers
| 120-55-8 | |
| 314.34 | |
| NXQMCAOPTPLPRL-UHFFFAOYSA-N | |
| 2-[2-(benzoyloxy)ethoxy]ethyl benzoate |
| C18H18O5 | |
| MFCD00020679 | |
| 8437 | |
| O=C(OCCOCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 120-55-8 | |
| 236°C | |
| MFCD00020679 | |
| NXQMCAOPTPLPRL-UHFFFAOYSA-N | |
| 2-[2-(benzoyloxy)ethoxy]ethyl benzoate | |
| 8437 | |
| ≥97.0% (GC) | |
| Diethylene Glycol Dibenzoate |
| Yellow | |
| C18H18O5 | |
| 25 mL | |
| O=C(OCCOCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 314.34 | |
| 314.34 | |
| Liquid |
Safety and Handling
RTECSNumber : ID6650000
TSCA : Yes