missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylene glycol mono-n-butyl ether, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A16068AU

 View more versions of this product

Catalog No. AAA16068AU


Explore available promotions

Description

Description

Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol mono-n-butyl ether has a wide variety of applications in Chiral chemistry and green chemistry. It is also used in cosmetics. It is used as diluents and leveling agents in the manufacture of paints and in baking. It is also used in the manufacture of nitrocellulose. In brake fluid, it is used as an additive. It is used in the printing industry due to its slow evaporation rate. It is also used as a fixative for perfumes and antiseptics. It is used as an additive to prevent ice buildup in jet fuel.

Solubility
Miscible with water, ethyl ether, ethanol acetone and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents and light metals.
Specifications

Specifications

Diethylene glycol mono-n-butyl ether
112-34-5
230°C to 232°C
100°C (212°F)
1.432
HOCH2CH2OCH2CH2O(CH2)3CH3
14,1557
Miscible with water,ethyl ether,ethanol acetone and benzene.
CCCCOCCOCCO
162.229
162.23
-68°C
0.967
1000 mL
Not Determined
C8H18O3
MFCD00002881
2-2-butoxyethoxy ethanol, diethylene glycol monobutyl ether, butyldiglycol, butyl carbitol, diethylene glycol butyl ether, butyl diglycol, butyl digol, ethanol, 2-2-butoxyethoxy, butoxyethoxyethanol, butoxydiglycol
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
2-(2-butoxyethoxy)ethanol
8177
99%

Chemical Identifiers

112-34-5
MFCD00002881
2-2-butoxyethoxy ethanol, diethylene glycol monobutyl ether, butyldiglycol, butyl carbitol, diethylene glycol butyl ether, butyl diglycol, butyl digol, ethanol, 2-2-butoxyethoxy, butoxyethoxyethanol, butoxydiglycol
CCCCOCCOCCO
C8H18O3
OAYXUHPQHDHDDZ-UHFFFAOYSA-N
8177
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P260-P264b-P280i-P305+P351+P338-P314-P501c

H319-H373

EINECSNumber : 203-961-6

RTECSNumber : KJ9100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only