missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylenetriamine, 98+%, Thermo Scientific Chemicals

Diethylenetriamine, 98+%, C4H13N3, CAS Number-111-40-0, 1,4,7-triazaheptane, 2,2'-diaminodiethylamine, diethylenetriamine, 2,2'-iminodiethylamine, bis 2-aminoethyl amine, barsamide 115, ancamine deta, n,n-bis 2-aminoethyl amine, epicure t, diethylene triamine, 2.5L, 98% min. (GC), 04,255

Supplier:  Thermo Scientific Chemicals 114312500

 View more versions of this product

Catalog No. AC114312500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylenetriamine
-40.0°C
206.0°C
Authentic
Glass bottle
1.4826 to 1.4846
MFCD00008171
04,255
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
RPNUMPOLZDHAAY-UHFFFAOYSA-N
N'-(2-aminoethyl)ethane-1,2-diamine
8111
CHEBI:30629
98+%
111-40-0
0.9600g/mL
96°C
98% min. (GC)
C4H13N3
H2NCH2CH2NHCH2CH2NH2
250 mL
0.96
Solubility in water: miscible. Other solubilities: soluble in most common organic solvents, soluble in hydrocarbons
NCCNCCN
103.17
5.05 mPa.s (20°C)
103.17
Liquid

Chemical Identifiers

111-40-0
103.17
RPNUMPOLZDHAAY-UHFFFAOYSA-N
8111
NCCNCCN
C4H13N3
MFCD00008171
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
CHEBI:30629
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth

GHS Signal Word: Danger

EINECSNumber : 203-865-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only