missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylenetriamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11033AP

 View more versions of this product

Catalog No. AAA11033AP


Explore available promotions

Description

Description

Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

Solubility
Miscible with water and ethanol. Immiscible with ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.
Specifications

Specifications

Diethylenetriamine
-40°C
200°C to 204°C
Ammonia-like
1.483
MFCD00008171
UN2079
Air Sensitive
Miscible with water and ethanol. Immiscible with ether.
NCCNCCN
103.17
CHEBI:30629
99%
111-40-0
0.952
94°C (201°F)
C4H13N3
H2NCH2CH2NHCH2CH2NH2
500 mL
605314
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
RPNUMPOLZDHAAY-UHFFFAOYSA-N
N'-(2-aminoethyl)ethane-1,2-diamine
8111
103.17

Chemical Identifiers

111-40-0
103.17
RPNUMPOLZDHAAY-UHFFFAOYSA-N
8111
N'-(2-aminoethyl)ethane-1,2-diamine
C4H13N3
MFCD00008171
diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
CHEBI:30629
NCCNCCN
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

P210-P235-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H302+H312-H314-H317-H330-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DIETHYLENETRIAMINE

EINECSNumber : 203-865-4

RTECSNumber : IE1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only