Learn More
Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1517803
Description
Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is-20°C. It is sensitive to moisture. Store under dry inert gas.
Specifications
Diethylenetriaminepentaacetic acid dianhydride | |
∽190°C (decomposition) | |
MFCD00010697 | |
1230954 | |
dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid | |
RAZLJUXJEOEYAM-UHFFFAOYSA-N | |
2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid | |
100825 | |
95% |
23911-26-4 | |
C14H19N3O8 | |
1 g | |
Moisture sensitive | |
Hydrolyzes in water. | |
C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O | |
357.319 | |
357.32 |
Chemical Identifiers
23911-26-4 | |
357.319 | |
RAZLJUXJEOEYAM-UHFFFAOYSA-N | |
100825 | |
C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O |
C14H19N3O8 | |
MFCD00010697 | |
dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid | |
2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only