missing translation for 'onlineSavingsMsg'
Learn More

Diethylsilane, 96+%

CAS: 542-91-6 | C4H10Si | 86.209 g/mol

$117.40 - $404.26

Chemical Identifiers

CAS 542-91-6
Molecular Formula C4H10Si
Molecular Weight (g/mol) 86.209
MDL Number MFCD00039877
InChI Key KZZFGAYUBYCTNX-UHFFFAOYSA-N
Synonym diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min
PubChem CID 6327565
IUPAC Name diethylsilicon
SMILES CC[Si]CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1646806
View Documents
Thermo Scientific Chemicals
L1646806
5 g
Each for $117.40
Only null left
 
AAL1646814
View Documents
Thermo Scientific Chemicals
L1646814
25 g
Each for $404.26
Only null left
 
Description

Description

Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.

Solubility
Miscible with terahydrofuran, diethyl ether, 1,4-dioxane and benzene.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, oxygen and water.
Specifications

Chemical Identifiers

542-91-6
86.209
KZZFGAYUBYCTNX-UHFFFAOYSA-N
6327565
CC[Si]CC
C4H10Si
MFCD00039877
diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min
diethylsilicon

Specifications

56°C
542-91-6
C4H10Si
UN1993
−20°C (−4°F)
diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min
KZZFGAYUBYCTNX-UHFFFAOYSA-N
diethylsilicon
6327565
≥98%
1.392
0.684
-132°C
5 g
MFCD00039877
1730902
14,3128
Miscible with terahydrofuran,diethyl ether,1,4-dioxane and benzene.
CC[Si]CC
86.209
88.23
Mild
Moisture sensitive
Diethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 208-834-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only