Learn More
Diethylvinylphosphonate, 97%, Thermo Scientific Chemicals
$268.30 - $268.30
Chemical Identifiers
CAS | 682-30-4 |
---|---|
Molecular Formula | C6H13O3P |
Molecular Weight (g/mol) | 164.14 |
MDL Number | MFCD00009079 |
InChI Key | DREPONDJUKIQLX-UHFFFAOYSA-N |
Synonym | diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g |
PubChem CID | 69629 |
SMILES | CCOP(=O)(OCC)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1036809
|
Thermo Scientific Chemicals
01036809 |
10 g |
Each for $268.30
|
|
|||||
Description
Diethylvinylphosphonate is used in the preparation of diethyl N-alkyl-2-aminoethylphosphonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
682-30-4 | |
164.14 | |
DREPONDJUKIQLX-UHFFFAOYSA-N | |
69629 |
C6H13O3P | |
MFCD00009079 | |
diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g | |
CCOP(=O)(OCC)C=C |
Specifications
682-30-4 | |
202°C | |
C6H13O3P | |
MFCD00009079 | |
UN3278 | |
diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g | |
CCOP(=O)(OCC)C=C | |
164.14 | |
164.14 | |
Liquid |
1.068 g/mL | |
110°C (230°F) | |
1.429 | |
10 g | |
EM>5 | |
DREPONDJUKIQLX-UHFFFAOYSA-N | |
1-[ethenyl(ethoxy)phosphoryl]oxyethane | |
69629 | |
97% | |
Diethylvinylphosphonate |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
EINECSNumber : 211-663-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only