missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Chromogenic Dehydrated Culture Media: MI Agar
Culture media tests drinking water for the presence of E.coli and total coliforms with membrane filtration method
$3121.58 - $3121.58
Specifications
| Color | Beige |
|---|---|
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | MI Agar |
Description
Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.
- Chromogenic-fluorogenic medium for detecting total coliforms and Escherichia coli in 24 hours or less
- Under ambient light, E. coli exhibits bluish-gray appearance; under long-wave UV light, it displays blue-green fluorescence
- Coliform are cream under ambient light, and display blue-white fluorescence under long-wave UV light
- Conforms to EPA Method 1604 for monitoring drinking water under the Total Coliform Rule, and source water under the Surface Water Treatment
Applications:
For testing ambient and potable water, treatment plant effluents, and beverage and pharmaceutical manufacturing
Order Info
Available in dehydrated form.
Specifications
| Beige | |
| Solid | |
| United States | |
| MI Agar |