missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Chromogenic Dehydrated Media: m-EI Agar Base
Culture media tests water for the presence of Enterococci by membrane filtration method
$1694.39 - $1694.39
Specifications
Content And Storage | 5°C to 25°C |
---|---|
Form | Solid |
Manufacturing Origin | United States |
Media Type | m EI Agar |
Formulation | Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L |
Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Certifications/Compliance | Price | Quantity | ||||
B14881
|
BD
214881 |
500 g | ISO/DIS, 11014 |
Each for $1,694.39
|
|
||||
Description
- Any colonies present will exhibit blue halo within 24 hours of the test
- Conforms to EPA Method 1600
Order Info
Available in dehydrated form.
Specifications
5°C to 25°C | |
United States | |
Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L |
Solid | |
m EI Agar | |
Bottle |