missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium
For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles
$241.55 - $1093.18
Specifications
Certifications/Compliance | AOAC, BAM, BS12, COMPF, EP, USDA, USP |
---|---|
Content And Storage | 2°C to 8°C |
Form | Dehydrated |
Media Type | Fluid Thioglycollate Medium |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Media Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Media Type | Price | Quantity | |||
DF0256-17-2
|
BD
225650 |
500 g | Bottle | Fluid Thioglycollate Medium |
Each for $241.55
|
|
|||
DF0256-07-4
|
BD
225620 |
2000 g | Pail | Fluid Thioglycollate Medium |
Each for $1,093.18
|
|
|||
Description
- Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Certifications
AOAC, BAM, BS12, COMPF, EP, USDA, USP
Specifications
AOAC, BAM, BS12, COMPF, EP, USDA, USP | |
Dehydrated | |
Microbiology Media | |
Used for sterility testing |
2°C to 8°C | |
Fluid Thioglycollate Medium | |
Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L |