missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: MacConkey Agar
For differential isolation of enteric bacilli based on lactose fermentation
$959.82 - $959.82
Specifications
Content And Storage | Refrigerate |
---|---|
Form | Dehydrated |
Media Type | MacConkey Agar |
Product Type | Agar |
Formulation | Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Certifications/Compliance | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Certifications/Compliance | Price | Quantity | |||
DF0075-07-3
|
BD
212122 |
2 kg | Pail | CE, SMD, SMWW, BS12, AOAC, BAM, COMPF, CCAM, JP, MCM9, EP, JP, MCM9 |
Each for $959.82
|
|
|||
Specifications
Refrigerate | |
MacConkey Agar | |
Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
Dehydrated | |
Agar | |
For differential isolation of enteric Bacilli based on Lactose fermentation |