missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Difco™ Dehydrated Culture Media: Membrane Fecal Coliform (mFC) Agar
For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures
$406.50 - $406.50
Specifications
Certifications/Compliance | AOAC, CCAM, EPA, SMWW |
---|---|
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | mFC Agar |
Product Type | Agar |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
DF0677-17-3
![]() |
BD
267720 |
500 g |
Each for $406.50
|
![]() |
||||||
Description
- Use with Rosolic Acid
- Formulation: Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L
Certifications
AOAC, CCAM, EPA, SMWW
Specifications
AOAC, CCAM, EPA, SMWW | |
Dehydrated | |
Agar | |
For Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperatures |
Refrigerate | |
mFC Agar | |
Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L | |
Bottle |