BD Difco™ Disposable Inoculating Needles and Loops
Sterile, disposable inoculating needles do not require flaming
$175.79 - $771.05
Catalog Number | Mfr. No. | Color | Volume | Quantity | Product Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Volume | Quantity | Product Type | Price | Quantity | ||
22-031-20
|
BD
220214 |
Light Green | 1 μL | 250/Pk. | Inoculating Loop |
Pack of 250 Each for $247.24
|
|
||
22-031-24
|
BD
220218 |
Violet | 1 μL | 1000/Pk. | Inoculating Needle |
Pack of 1000 Each for $492.61
|
|
||
22-031-21
|
BD
220215 |
Light Green | 1 μL | 1000/Pk. | Inoculating Loop |
Pack of 1000 Each for $771.05
|
|
||
22-031-22
|
BD
220216 |
Light Blue | 10 μL | 250/Pk. | Inoculating Loop |
Pack of 250 Each for $175.79
|
|
||
22-031-23
|
BD
220217 |
Light Blue | 10 μL | 1000/Pk. | Inoculating Loop |
Pack of 1000 Each for $608.87
|
|
||
Description
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifications
Plastic Resin | |
Sterile | |
Single-use |