Learn More
Diffraction Viewing Glasses
See the spectrum with these specs.
Supplier: Customer Channels Group (Ccg) S48814
Description
- Cardboard glasses use a small, flexible, diffraction grating that breaks up light into the colors of the spectrum
- Great for classroom spectroscopy experiments and elementary physical science investigations
- Individually sold glasses have green frames; packs have assorted color frames
Spécifications
| Diffraction Viewing Glass | |
| Great for classroom spectroscopy experiments and elementary physical science investigations | |
| 1 green frame |