Diffraction Viewing Glasses
See the spectrum with these specs.
Supplier:  Customer Channels Group (Ccg) S48814A
 View more versions of this product
Catalog No. S48814A
Explore available promotions
Description
Description
  • Cardboard glasses use a small, flexible, diffraction grating that breaks up light into the colors of the spectrum
  • Great for classroom spectroscopy experiments and elementary physical science investigations
  • Individually sold glasses have green frames; packs have assorted color frames
Specifications
Specifications
Diffraction Viewing Glass
Great for classroom spectroscopy experiments and elementary physical science investigations
10 assorted color frames