missing translation for 'onlineSavingsMsg'
Learn More

Difloxacin hydrochloride

CAS: 91296-86-5 | C21H20ClF2N3O3 | 435.856 g/mol

$263.03 - $863.23

Chemical Identifiers

CAS 91296-86-5
Molecular Formula C21H20ClF2N3O3
Molecular Weight (g/mol) 435.856
MDL Number MFCD03840489
InChI Key JFMGBGLSDVIOHL-UHFFFAOYSA-N
Synonym difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
PubChem CID 56205
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6375406
View Documents
Thermo Scientific Chemicals
J6375406
5 g
Each for $263.03
Only null left
 
AAJ6375414
View Documents
Thermo Scientific Chemicals
J6375414
25 g
Each for $863.23
Only null left
 
Description

Description

A quinolone antibacterial compound. Structurally related to norfloxacin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A quinolone antibacterial compound
  • Structurally related to norfloxacin
Specifications

Chemical Identifiers

91296-86-5
435.856
JFMGBGLSDVIOHL-UHFFFAOYSA-N
56205
CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl
C21H20ClF2N3O3
MFCD03840489
difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride

Specifications

91296-86-5
C21H20ClF2N3O3
5 g
14,3141
Soluble in aqueous buffer
CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl
435.856
435.85
Difloxacin hydrochloride
White
MFCD03840489
6464798
difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
JFMGBGLSDVIOHL-UHFFFAOYSA-N
6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
56205
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only