missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Digi-Sense™ Precalibrated Pocket Size Moisture Meter

Rugged pocket sized meter with one-handed operation

Supplier:  Digi-Sense™ 2025035

Catalog No. 13-245-286


Explore available promotions

Includes: Four LR44 batteries and NIST-traceable calibration report with data supplied by InnoCal.

Description

Description

  • Reliable measurements guaranteed
  • Precalibrated to NIST-traceable standards
  • Measure the moisture level in wood and hardened materials
  • Low battery indication

Warranty

2 Years

Specifications

Specifications

Four LR44 batteries and NIST-traceable calibration report with data supplied by InnoCal.
2 cm
4 cm
Wood: ±2%; Material: ±0.2%; Temp: ±1.8° F (1°C)
Moisture Meter
9 cm
Videos
SDS
Documents

Documents

Promotions

Promotions