Digi-Sense™ Temp-16 Single-Input RTD Thermometer
Heavy-duty Digi-Sense™ Temp-16 Single-Input RTD Thermometer offers versatility and accuracy in a compact design
Supplier:  Digi-Sense™ 9142807
Catalog No. 13-300-205
Explore available promotions
Description
Description
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
Specifications
Specifications
CE
Protective rubber armor/boot with stand and three AA batteries.
None
1562°F
-328°F
1.5 in. Display, 4-digit LCD
No
0.1°; 1° below -148°F (-200°C)
Temp16 Single-Input RTD Thermometer
850°C
-200°C