missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Digital Dry Baths/Block Heaters

Increase lab versatility with the Thermo Scientific™ digital dry baths/block heaters.

$1102.09 - $1575.41

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID digital
Lighting LED
Material Powder-coated Steel
Relative Humidity <80%
View More Specs

Products 3
Catalog Number Mfr. No. No. of Blocks Heating Rate Voltage Includes Price Quantity  
Catalog Number Mfr. No. No. of Blocks Heating Rate Voltage Includes Price Quantity  
88870001
Accessories View Documents
Thermo Scientific™
88870001
1 (order separately) ≤ 20 min. 30° to 130°C 100/120 V Dry bath, US style plug, block lifter
Each for $1,177.71
 
88870002
Accessories View Documents
Thermo Scientific™
88870002
2 (order separately) ≤ 20 min. 30° to 130°C 100/120 V Dry bath, US style plug, block lifter
Each for $1,102.09
 
88870003
Accessories View Documents
Thermo Scientific™
88870003
4 (order separately) ≤ 25 min. 30° to 130°C 100/120 V Dry bath, US style plug, block lifter
Each for $1,575.41
 
Description

Description

Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration allows the user to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device ensures sample and user safety
  • Temperature deviation adjustment
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required Accessory:
  • Unit must be ordered with block(s)
    Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
    • Digital controls and display of time and temperature
    • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
    • Precise temperature control with PID circuit
    • Temperature calibration allows the user to offset the temperature to desired value
    • Timer allows the user to accurately monitor the heating time
    • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
    • Powder-coated steel body construction ensures durability
    • Built-in over-temperature protection device ensures sample and user safety
    • Temperature deviation adjustment
    Applications:
    • Sample preparation
    • Enzyme preservation
    • Enzyme-substrate reactions
    • DNA amplification
    • Electrophoresis gel degeneration
    • Serum coagulation
    Required Accessory:
    • Unit must be ordered with block(s)

      Warranty

      2 Year, Replacement

Specifications

Specifications

CE, cULus, RoHS
LED
<80%
Dry Bath/Block
≤ ± 1°C
2 Year, Replacement
200 mm
100 mm
PID digital
Powder-coated Steel
Ambient +5° to 130°C (Ambient at 25°C)
≤ ± 0.5°C
0 to 99:59 min or continuous
7.9 in.
3.9 in.
50/60 Hz
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.