Stoelting™ Digital Ultrasonic Cleaner
Stoelting™ Digital Ultrasonic Cleaner is a high efficiency cleaner specially developed to clean surgical instruments.
Supplier: Stoelting™ STG58620
Description
- Ultrasonic waves pump micron-sized bubbles into the water.
- The bubbles fit into the tightest spaces that would be impossible to clean by traditional methods.
- When the bubbles implode, water hits the instrument at speeds up to 250 mph-a very powerful cleaning action.
- Only available in 110 V
Specifications
Neuroscience | |
Ultrasonic |