missing translation for 'onlineSavingsMsg'
Learn More

Diglycerol 80.0+%, TCI America™

$65.24 - $208.78

Chemical Identifiers

CAS 627-82-7
Molecular Formula C6H14O5
Molecular Weight (g/mol) 166.173
MDL Number MFCD00049316
InChI Key GPLRAVKSCUXZTP-UHFFFAOYSA-N
Synonym diglycerol, diglycerine, diglycerin, alpha,alpha'-diglycerol, glyceryl ether, 1,2-propanediol, 3,3'-oxybis, oxydi propanediol, tetrahydroxydipropyl ether, .alpha.,.alpha.'-diglycerol, 3-2,3-dihydroxypropoxy propane-1,2-diol
PubChem CID 42953
IUPAC Name 3-(2,3-dihydroxypropoxy)propane-1,2-diol
SMILES C(C(COCC(CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T011925G
View Documents
TCI America
T011925G
25 g
Each for $65.24
Only null left
 
T0119500G
View Documents
TCI America
T0119500G
500 g
Each for $208.78
Only null left
 
Description

Description

Specifications

Chemical Identifiers

627-82-7
166.173
GPLRAVKSCUXZTP-UHFFFAOYSA-N
42953
C(C(COCC(CO)O)O)O
C6H14O5
MFCD00049316
diglycerol, diglycerine, diglycerin, alpha,alpha'-diglycerol, glyceryl ether, 1,2-propanediol, 3,3'-oxybis, oxydi propanediol, tetrahydroxydipropyl ether, .alpha.,.alpha.'-diglycerol, 3-2,3-dihydroxypropoxy propane-1,2-diol
3-(2,3-dihydroxypropoxy)propane-1,2-diol

Specifications

627-82-7
C6H14O5
25 g
GPLRAVKSCUXZTP-UHFFFAOYSA-N
3-(2,3-dihydroxypropoxy)propane-1,2-diol
42953
≥80.0% (GC)
Diglycerol
Yellow
MFCD00049316
diglycerol, diglycerine, diglycerin, alpha,alpha'-diglycerol, glyceryl ether, 1,2-propanediol, 3,3'-oxybis, oxydi propanediol, tetrahydroxydipropyl ether, .alpha.,.alpha.'-diglycerol, 3-2,3-dihydroxypropoxy propane-1,2-diol
C(C(COCC(CO)O)O)O
166.173
166.17
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0418

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'