missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diglycolic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 162265000

 View more versions of this product

Catalog No. AC162265000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diglycolic acid
141.0°C to 145.0°C
White to Beige
C4H6O5
MFCD00004309
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
QEVGZEDELICMKH-UHFFFAOYSA-N
2-(carboxymethoxy)acetic acid
8088
98%
Authentic
Crystalline Powder
98%
110-99-6
98%
O(CH2CO2H)2
03, 234
Solubility in water: soluble. Other solubilities: soluble in ethanol
OC(=O)COCC(O)=O
134.09
134.09
500 g
Plastic bottle

Chemical Identifiers

110-99-6
134.09
QEVGZEDELICMKH-UHFFFAOYSA-N
8088
OC(=O)COCC(O)=O
C4H6O5
MFCD00004309
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
2-(carboxymethoxy)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-823-5

RTECSNumber : AJ1530000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only