Learn More
Digoxin, 96%, Thermo Scientific Chemicals
$253.84 - $858.30
Chemical Identifiers
CAS | 20830-75-5 |
---|---|
Molecular Formula | C41H64O14 |
Molecular Weight (g/mol) | 780.949 |
MDL Number | MFCD00003674 |
InChI Key | LTMHDMANZUZIPE-PUGKRICDSA-N |
Synonym | digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn |
PubChem CID | 2724385 |
ChEBI | CHEBI:4551 |
IUPAC Name | 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet |
SMILES | CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2190203
|
Thermo Scientific Chemicals
B2190203 |
1 g |
Each for $253.84
|
|
||||||
AAB2190206
|
Thermo Scientific Chemicals
B2190206 |
5 g |
Each for $858.30
|
|
||||||
Description
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDigoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
20830-75-5 | |
780.949 | |
LTMHDMANZUZIPE-PUGKRICDSA-N | |
2724385 | |
3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet |
C41H64O14 | |
MFCD00003674 | |
digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn | |
CHEBI:4551 | |
CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O |
Specifications
20830-75-5 | |
C41H64O14 | |
1 g | |
14,3167 | |
Soluble in dimethyl sulfoxide and methanol. | |
CC1C(C(CC(O1)OC2C(OC(CC2O)OC3C(OC(CC3O)OC4CCC5(C(C4)CCC6C5CC(C7(C6(CCC7C8=CC(=O)OC8)O)C)O)C)C)C)O)O | |
780.949 | |
CHEBI:4551 | |
96% |
∼250°C (decomposition) | |
MFCD00003674 | |
UN2811 | |
digoxin, 12beta-hydroxydigitoxin, digoxine, lanoxin, lanoxicaps, digosin, digossina, digoxina, digoxinum, lanoxicaps tn | |
LTMHDMANZUZIPE-PUGKRICDSA-N | |
3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tet | |
2724385 | |
780.95 | |
Digoxin |
Safety and Handling
GHS H Statement
H300
Fatal if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
H300-H336-H351-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 244-068-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only