missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, Premion™, 99.95% (metals basis), Pt 37.5% min

CAS: 18497-13-7 | Cl6H14O6Pt | 517.89 g/mol

$266.56 - $5246.49

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3625903
View Documents
Thermo Scientific Chemicals
03625903
1 g
Each for $266.56
Only null left
 
AA3625906
View Documents
Thermo Scientific Chemicals
03625906
5 g
Each for $1,145.68
Only null left
 
AA3625914
View Documents
Thermo Scientific Chemicals
03625914
25 g
Each for $5,246.49
Only null left
 
Description

Description

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18497-13-7
517.89
PIJUVEPNGATXOD-UHFFFAOYSA-J
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
Cl6H14O6Pt
MFCD00149910
platinum(4+) dihydrogen hexahydrate hexachloride

Specifications

60°C (decomposition)
Red to Brown
1 g
Cl6H14O6Pt
Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
517.89
ACS
Red-brown lump
Dihydrogen hexachloroplatinate(IV) hexahydrate
18497-13-7
Lump
99.95% (metals basis)
MFCD00149910
PIJUVEPNGATXOD-UHFFFAOYSA-J
platinum(4+) dihydrogen hexahydrate hexachloride
517.91 (409.82 Anhydrous)
Hygroscopic
2.43
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301-H314-H317-H334-H335

DOTInformation : DOT Class: 8; Packing Group: III

EINECSNumber : 241-010-7

RTECSNumber : TP1500000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only