missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dihydrogen hexafluorozirconate, 20% w/w aq. soln., Thermo Scientific Chemicals

$129.47 - $129.47

Chemical Identifiers

CAS 12021-95-3
Molecular Formula F6H2Zr
Molecular Weight (g/mol) 207.23
MDL Number MFCD00082965
InChI Key PZFQYBOZXGWNAM-UHFFFAOYSA-H
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39461A1
View Documents
Thermo Scientific Chemicals
039461A1
1 kg
Each for $129.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12021-95-3
207.23
PZFQYBOZXGWNAM-UHFFFAOYSA-H
F6H2Zr
MFCD00082965

Specifications

12021-95-3
1.52 g/mL
MFCD00082965
UN3264
207.23
20% w/w aq. soln.
Dihydrogen hexafluorozirconate
Green
F6H2Zr
1 kg
PZFQYBOZXGWNAM-UHFFFAOYSA-H
207.22
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302+H312-H314-H331-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 234-666-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only