missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dihydrogen hexafluorozirconate, 45% w/w aq. soln., Thermo Scientific Chemicals

$332.44 - $332.44

Chemical Identifiers

CAS 12021-95-3
Molecular Formula F6H2Zr
Molecular Weight (g/mol) 207.23
MDL Number MFCD00082965
InChI Key PZFQYBOZXGWNAM-UHFFFAOYSA-H
Synonym Hexafluorozirconic acid; Hydrogen hexafluorozirconate
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39462A3
View Documents
Thermo Scientific Chemicals
039462A3
2 kg
Each for $332.44
 
Description

Description

Dihydrogen hexafluorozirconate is used in manufacturing of optical glass and fluozirconate. It acts as a precursor to zirconium dioxide ceramic thin films. Further, it is used for surface pretreatments on steel to prevent corrosion. In addition to this, it is used in the production of the atomic energy industry, advanced electrical materials and refractories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12021-95-3
207.23
PZFQYBOZXGWNAM-UHFFFAOYSA-H
F6H2Zr
MFCD00082965
Hexafluorozirconic acid; Hydrogen hexafluorozirconate

Specifications

12021-95-3
1.51 g/mL
MFCD00082965
UN2922
Miscible with acid-base solutions.
207.23
45% w/w aq. soln.
Dihydrogen hexafluorozirconate
Colorless
F6H2Zr
2 kg
Hexafluorozirconic acid; Hydrogen hexafluorozirconate
PZFQYBOZXGWNAM-UHFFFAOYSA-H
207.22
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H301+H311+H331-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 234-666-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only