missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

$79.87 - $2213.52

Chemical Identifiers

CAS 75-11-6
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169830250
SDS View Documents
Thermo Scientific Chemicals
169830250
25 g Glass bottle
Each for $79.87
 
AC169831000
SDS View Documents
Thermo Scientific Chemicals
169831000
100 g Glass bottle
Each for $215.38
 
AC169835000
SDS View Documents
Thermo Scientific Chemicals
169835000
500 g Glass bottle
Each for $677.47
 
AC169830025
SDS View Documents
Thermo Scientific Chemicals
169830025
2.5 kg Glass Bottle
Each for $2,213.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-11-6
267.84
NZZFYRREKKOMAT-UHFFFAOYSA-N
6346
ICI
CH2I2
MFCD00001079
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
diiodomethane

Specifications

75-11-6,7440-22-4
Red to Yellow
181.0°C
Authentic
Glass bottle
1.737
MFCD00001079
01,71
3.32
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
NZZFYRREKKOMAT-UHFFFAOYSA-N
diiodomethane
Silver wire
2.80 mPa.s (20°C)
99+%
Diiodomethane
6.0°C
3.3200g/mL
>110°C
99% min. (GC)
CH2I2
CH2I2
25 g
09,170; 11,182; 17,116
15,6137
Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene
ICI
267.84
6346
267.83
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 200-841-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only