missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyl Phosphite 95.0+%, TCI America™
$29.83 - $268.43
Chemical Identifiers
| CAS | 1189-24-8 |
|---|---|
| Molecular Formula | C8H18O3P+ |
| Molecular Weight (g/mol) | 193.203 |
| InChI Key | QPKOILOWXGLVJS-UHFFFAOYSA-N |
| Synonym | Phosphorous Acid Diisobutyl Ester, Diisobutyl Phosphonate, Phosphonic Acid Diisobutyl Ester |
| PubChem CID | 6327151 |
| IUPAC Name | bis(2-methylpropoxy)-oxophosphanium |
| SMILES | CC(C)CO[P+](=O)OCC(C)C |
Chemical Identifiers
| 1189-24-8 | |
| 193.203 | |
| Phosphorous Acid Diisobutyl Ester, Diisobutyl Phosphonate, Phosphonic Acid Diisobutyl Ester | |
| bis(2-methylpropoxy)-oxophosphanium |
| C8H18O3P+ | |
| QPKOILOWXGLVJS-UHFFFAOYSA-N | |
| 6327151 | |
| CC(C)CO[P+](=O)OCC(C)C |
Specifications
| 1189-24-8 | |
| 138°C | |
| 25 g | |
| QPKOILOWXGLVJS-UHFFFAOYSA-N | |
| bis(2-methylpropoxy)-oxophosphanium | |
| 6327151 | |
| ≥95.0% (GC) | |
| Diisobutyl Phosphite |
| Yellow | |
| C8H18O3P+ | |
| Phosphorous Acid Diisobutyl Ester, Diisobutyl Phosphonate, Phosphonic Acid Diisobutyl Ester | |
| CC(C)CO[P+](=O)OCC(C)C | |
| 193.203 | |
| 194.21 | |
| Liquid |
Safety and Handling
TSCA : No