missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ketone, 98%

CAS: 565-80-0 | C7H14O | 114.188 g/mol

$76.79 - $278.92

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1287522
View Documents
Thermo Scientific Chemicals
A1287522
100 g
Each for $76.79
Only null left
 
AAA1287536
View Documents
Thermo Scientific Chemicals
A1287536
500 g
Each for $278.92
Only null left
 
Description

Description

2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
Specifications

Chemical Identifiers

565-80-0
114.188
HXVNBWAKAOHACI-UHFFFAOYSA-N
11271
CC(C)C(=O)C(C)C
C7H14O
MFCD00008918
2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
2,4-dimethylpentan-3-one

Specifications

565-80-0
0.803
15°C (59°F)
1.399
100 g
773782
Not miscible or difficult to mix in water. Soluble in alcohol.
CC(C)C(=O)C(C)C
114.188
114.19
Diisopropyl ketone
-69°C
124°C to 125°C
C7H14O
MFCD00008918
UN1224
2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
HXVNBWAKAOHACI-UHFFFAOYSA-N
2,4-dimethylpentan-3-one
11271
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H225-H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 209-294-7

RTECSNumber : SA8575500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only