missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-Diisopropyl L-tartrate, 97%, Thermo Scientific Chemicals

$90.22 - $290.36

Chemical Identifiers

CAS 2217-15-4
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.248
MDL Number MFCD00064450
InChI Key XEBCWEDRGPSHQH-HTQZYQBOSA-N
Synonym +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 6453580
IUPAC Name dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1694114
View Documents
Thermo Scientific Chemicals
A1694114
25 g
Each for $90.22
 
AAA1694122
View Documents
Thermo Scientific Chemicals
A1694122
100 g
Each for $290.36
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2217-15-4
234.248
XEBCWEDRGPSHQH-HTQZYQBOSA-N
6453580
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
C10H18O6
MFCD00064450
+-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate

Specifications

2217-15-4
150°C to 151°C (11 mmHg)
C10H18O6
MFCD00064450
1727863
XEBCWEDRGPSHQH-HTQZYQBOSA-N
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
234.248
234.25
(+)-Diisopropyl L-tartrate
1.117
110°C (230°F)
1.44
25 g
+-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
+16° (Neat)
dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
6453580
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-709-0

RTECSNumber : WW8100000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only