missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl Sulfate 97.0+%, TCI America™
$268.43 - $1083.02
Chemical Identifiers
| CAS | 2973-10-6 |
|---|---|
| Molecular Formula | C6H14O4S |
| Molecular Weight (g/mol) | 182.23 |
| MDL Number | MFCD01682898 |
| InChI Key | HWBLTYHIEYOAOL-UHFFFAOYSA-N |
| PubChem CID | 18096 |
| ChEBI | CHEBI:82320 |
| IUPAC Name | dipropan-2-yl sulfate |
| SMILES | CC(C)OS(=O)(=O)OC(C)C |
Chemical Identifiers
| 2973-10-6 | |
| 182.23 | |
| HWBLTYHIEYOAOL-UHFFFAOYSA-N | |
| CHEBI:82320 | |
| CC(C)OS(=O)(=O)OC(C)C |
| C6H14O4S | |
| MFCD01682898 | |
| 18096 | |
| dipropan-2-yl sulfate |
Specifications
| 2973-10-6 | |
| Yellow | |
| C6H14O4S | |
| 5 g | |
| HWBLTYHIEYOAOL-UHFFFAOYSA-N | |
| dipropan-2-yl sulfate | |
| 18096 | |
| 182.23 | |
| Liquid |
| -19°C | |
| 94°C | |
| MFCD01682898 | |
| 1760 | |
| CC(C)OS(=O)(=O)OC(C)C | |
| 182.23 | |
| CHEBI:82320 | |
| ≥97.0% (GC) | |
| Diisopropyl Sulfate |
Safety and Handling
EINECSNumber : (2)-1673
RTECSNumber : WS8050000
TSCA : No
Recommended Storage : Freezer