Learn More
Diisopropylamine dichloroacetate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H61341MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diisopropylamine dichloroacetate | |
White | |
[(CH3)2CH]2NH·Cl2CHCO2H | |
250 mg | |
ILKBHIBYKSHTKQ-UHFFFAOYSA-N | |
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine | |
12617 | |
95% |
660-27-5 | |
C8H17Cl2NO2 | |
MFCD00868288 | |
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam | |
OC(=O)C(Cl)Cl.CC(C)NC(C)C | |
230.13 | |
230.13 | |
Crystalline Powder |
Chemical Identifiers
660-27-5 | |
230.13 | |
ILKBHIBYKSHTKQ-UHFFFAOYSA-N | |
12617 | |
OC(=O)C(Cl)Cl.CC(C)NC(C)C |
C8H17Cl2NO2 | |
MFCD00868288 | |
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam | |
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 211-538-2
RTECSNumber : AG6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only