missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diisopropylamine dichloroacetate, 95%, Thermo Scientific Chemicals

$74.67 - $797.92

Chemical Identifiers

CAS 660-27-5
Molecular Formula C8H17Cl2NO2
Molecular Weight (g/mol) 230.13
MDL Number MFCD00868288
InChI Key ILKBHIBYKSHTKQ-UHFFFAOYSA-N
Synonym diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
PubChem CID 12617
IUPAC Name 2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
SMILES OC(=O)C(Cl)Cl.CC(C)NC(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61341MD
View Documents
Thermo Scientific Chemicals
H61341MD
250 mg
Each for $74.67
 
AAH6134103
View Documents
Thermo Scientific Chemicals
H6134103
1 g
Each for $213.51
 
AAH6134106
View Documents
Thermo Scientific Chemicals
H6134106
5 g
Each for $797.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

660-27-5
230.13
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
12617
OC(=O)C(Cl)Cl.CC(C)NC(C)C
C8H17Cl2NO2
MFCD00868288
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine

Specifications

660-27-5
C8H17Cl2NO2
MFCD00868288
diisopropylamine dichloroacetate, diisopropylammonium dichloroacetate, diisopropylamine 2,2-dichloroacetate, vasculopatina, dipromonium, cubisol, dapocel, disotat, kalodil, oxypangam
OC(=O)C(Cl)Cl.CC(C)NC(C)C
230.13
230.13
Crystalline Powder
White
[(CH3)2CH]2NH·Cl2CHCO2H
250 mg
ILKBHIBYKSHTKQ-UHFFFAOYSA-N
2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
12617
95%
Diisopropylamine dichloroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 211-538-2

RTECSNumber : AG6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only