Learn More
Dilauroyl peroxide, 99%, Thermo Scientific Chemicals
$109.57 - $325.00
Chemical Identifiers
CAS | 105-74-8 |
---|---|
Molecular Formula | C24H46O4 |
Molecular Weight (g/mol) | 398.63 |
MDL Number | MFCD00008964 |
InChI Key | YIVJZNGAASQVEM-UHFFFAOYSA-N |
Synonym | lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle |
PubChem CID | 7773 |
ChEBI | CHEBI:82479 |
IUPAC Name | dodecanoyl dodecaneperoxoate |
SMILES | CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC349741000
|
Thermo Scientific Chemicals
349741000 |
100 g | Plastic bottle |
Each for $109.57
|
|
|||||
AC349745000
|
Thermo Scientific Chemicals
349745000 |
500 g | Plastic bottle |
Each for $325.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
105-74-8 | |
398.63 | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
7773 | |
dodecanoyl dodecaneperoxoate |
C24H46O4 | |
MFCD00008964 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
CHEBI:82479 | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Specifications
105-74-8 | |
White | |
98.5% min. (Iodometry) | |
C24H46O4 | |
MFCD00008964 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
dodecanoyl dodecaneperoxoate | |
7773 | |
398.63 | |
Powder |
53°C to 57°C | |
Authentic | |
Plastic bottle | |
[CH3(CH2)10CO]2O2 | |
100 g | |
Solubility in water: insoluble. Other solubilities: partly soluble in toluene and hexane | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC | |
398.63 | |
CHEBI:82479 | |
99% | |
Dilauroyl peroxide, 99% |
Safety and Handling
GHS H Statement
Heating may cause a fire.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig
GHS Signal Word: Danger
EINECSNumber : 203-326-3
RTECSNumber : OF2625000
TSCA : TSCA
RUO – Research Use Only