missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 115560010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethoxymethane | |
-105.0°C | |
0.8600g/mL | |
−18°C | |
99.5% min. (GC) | |
C3H8O2 | |
CH2(OCH3)2 | |
1 L | |
0.86 | |
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
dimethoxymethane | |
8020 | |
CHEBI:48341 | |
99.5+% |
109-87-5 | |
Colorless | |
41.0°C to 42.0°C | |
Authentic | |
Glass bottle | |
1.3525 to 1.3545 | |
MFCD00008495 | |
01,671; 02,271; 06,374; 07,115 | |
15, 6086 | |
Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene | |
COCOC | |
76.09 | |
3.25 mPa.s (20°C) | |
76.09 | |
Liquid |
Chemical Identifiers
109-87-5 | |
76.09 | |
NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
8020 | |
dimethoxymethane |
C3H8O2 | |
MFCD00008495 | |
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
CHEBI:48341 | |
COCOC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 203-714-2
RUO – Research Use Only