missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 115560025

 View more versions of this product

Catalog No. AC115560025



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethoxymethane
-105.0°C
0.8600g/mL
−18°C
99.5% min. (GC)
C3H8O2
CH2(OCH3)2
2.5 L
0.86
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
NKDDWNXOKDWJAK-UHFFFAOYSA-N
dimethoxymethane
8020
CHEBI:48341
99.5+%
109-87-5
Colorless
41.0°C to 42.0°C
Authentic
Glass bottle
1.3525 to 1.3545
MFCD00008495
01,671; 02,271; 06,374; 07,115
15, 6086
Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene
COCOC
76.09
3.25 mPa.s (20°C)
76.09
Liquid

Chemical Identifiers

109-87-5
76.09
NKDDWNXOKDWJAK-UHFFFAOYSA-N
8020
dimethoxymethane
C3H8O2
MFCD00008495
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
CHEBI:48341
COCOC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Danger

EINECSNumber : 203-714-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only