missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl (1-Diazo-2-oxopropyl)phosphonate 97.0+%, TCI America™
Supplier: TCI America D35465G
Specifications
Dimethyl (1-Diazo-2-oxopropyl)phosphonate | |
C5H9N2O4P | |
5 g | |
SQHSJJGGWYIFCD-UHFFFAOYSA-N | |
1-diazonio-1-dimethoxyphosphorylprop-1-en-2-olate | |
53397103 | |
≥97.0% (HPLC) |
90965-06-3 | |
MFCD07368360 | |
(1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Ohira-Bestmann Reagent | |
CC(=C([N+]#N)P(=O)(OC)OC)[O-] | |
192.111 | |
192.11 | |
Liquid |
Chemical Identifiers
90965-06-3 | |
192.111 | |
SQHSJJGGWYIFCD-UHFFFAOYSA-N | |
53397103 | |
CC(=C([N+]#N)P(=O)(OC)OC)[O-] |
C5H9N2O4P | |
MFCD07368360 | |
(1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Ohira-Bestmann Reagent | |
1-diazonio-1-dimethoxyphosphorylprop-1-en-2-olate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator